siRNA Details
| Gene Symbol: Gm16062 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045686.1 | 2 | 539 | |||
| GenBank mRNA | AK144909.1 | - | 2 | 506 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm16062| Gene Symbol: Gm16062 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045686.1 | 2 | 539 | |||
| GenBank mRNA | AK144909.1 | - | 2 | 506 | ||