siRNA Details
| Gene Symbol: Gm14379 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026741.1 | 2 | 697 | |||
| GenBank mRNA | AK040575.1 | - | 2 | 511 | ||
| AK155183.1 | - | 2 | 698 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB636260.1 | - | 2 | 508 | Chr. X - 7376114 - 7377014 [+] on Build GRCm38 | |

Gm14379
Genomic Map