siRNA Details
| Gene Symbol: 4930486F22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK133288.1 | - | 437 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB616668.1 | - | 3 | 437 | Chr. 10 - 86095839 - 86127120 [+] on Build GRCm38 | |

4930486F22Rik
Genomic Map