siRNA Details
| Gene Symbol: Gt(pU21)129Imeg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AB191465.1 | - | 1 | 134 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gt(pU21)129Imeg| Gene Symbol: Gt(pU21)129Imeg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AB191465.1 | - | 1 | 134 | ||