siRNA Details
| Gene Symbol: Gm12130 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040295.1 | 3 | 522 | |||
| GenBank mRNA | AK145334.1 | - | 3 | 492 | ||
| CJ052615.1 | - | 3 | 524 | |||
| DV652764.1 | - | 3 | 466 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | DV659262.1 | - | 4 | 605 | Chr. 11 - 38493336 - 38520002 [-] on Build GRCm38 | |

Gm12130
Genomic Map