siRNA Details
| Gene Symbol: 1810021B22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040417.1 | 4 | 594 | |||
| GenBank mRNA | AK007571.1 | - | 4 | 595 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1810021B22Rik| Gene Symbol: 1810021B22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040417.1 | 4 | 594 | |||
| GenBank mRNA | AK007571.1 | - | 4 | 595 | ||