siRNA Details
| Gene Symbol: Scpep1os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045955.1 | 3 | 374 | |||
| GenBank mRNA | AK015102.1 | - | 3 | 375 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY714635.1 | - | 3 | 375 | Chr. 11 - 88905928 - 88936015 [+] on Build GRCm38 | |

Scpep1os
Genomic Map