siRNA Details
| Gene Symbol: 0610040F04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_104577.1 | 7 | 1166 | |||
| GenBank mRNA | AK039720.1 | - | 7 | 1166 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

0610040F04Rik| Gene Symbol: 0610040F04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_104577.1 | 7 | 1166 | |||
| GenBank mRNA | AK039720.1 | - | 7 | 1166 | ||