siRNA Details
| Gene Symbol: 0610043K17Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040640.1 | 4 | 515 | |||
| GenBank mRNA | AK002929.1 | - | 4 | 516 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

0610043K17Rik| Gene Symbol: 0610043K17Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040640.1 | 4 | 515 | |||
| GenBank mRNA | AK002929.1 | - | 4 | 516 | ||