siRNA Details
| Gene Symbol: Gm13710 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046046.1 | 3 | 381 | |||
| GenBank mRNA | AK044106.1 | - | 3 | 382 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm13710| Gene Symbol: Gm13710 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046046.1 | 3 | 381 | |||
| GenBank mRNA | AK044106.1 | - | 3 | 382 | ||