siRNA Details
Gene Symbol: Gm15541 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK144575.1 | - | 2390 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gm15541 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK144575.1 | - | 2390 |