siRNA Details
| Gene Symbol: 1700023C21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045909.1 | 3 | 245 | |||
| GenBank mRNA | AK006264.1 | - | 3 | 249 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY706345.1 | - | 3 | 246 | Chr. 11 - 109845878 - 109848476 [+] on Build GRCm38 | |

1700023C21Rik
Genomic Map