siRNA Details
| Gene Symbol: 2310024H09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK009494.1 | - | 2 | 687 | ||
| Gene Symbol: Gm11266 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BY709204.1 | - | 2 | 687 | ||

2310024H09Rik
Genomic Map