siRNA Details
| Gene Symbol: A330041J22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045835.1 | 3 | 1262 | |||
| GenBank mRNA | AK039418.1 | - | 3 | 1245 | ||
| AK046134.1 | - | 3 | 1161 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | NM_176941.1 | - | 1245 | Chr. - - [-] on Build | ||

A330041J22Rik
Genomic Map