siRNA Details
Gene Symbol: 2900089D17Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK013828.1 | - | 2 | 187 | ||
AK019366.1 | - | 2 | 187 | |||
AK083239.1 | - | 2 | 2152 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 2900089D17Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK013828.1 | - | 2 | 187 | ||
AK019366.1 | - | 2 | 187 | |||
AK083239.1 | - | 2 | 2152 |