siRNA Details
| Gene Symbol: 2810001G20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033780.1 | 2 | 654 | |||
| GenBank mRNA | AK005116.1 | - | 2 | 621 | ||
| AK007927.1 | - | 2 | 656 | |||
| AK012631.1 | - | 2 | 647 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

2810001G20Rik| Gene Symbol: 2810001G20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033780.1 | 2 | 654 | |||
| GenBank mRNA | AK005116.1 | - | 2 | 621 | ||
| AK007927.1 | - | 2 | 656 | |||
| AK012631.1 | - | 2 | 647 | |||