siRNA Details
| Gene Symbol: Zmiz1os1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK047978.1 | - | 1464 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zmiz1os1| Gene Symbol: Zmiz1os1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK047978.1 | - | 1464 | |||