siRNA Details
| Gene Symbol: Gm11412 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK085025.1 | - | 1404 | |||
| Gene Symbol: LOC102635467 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK139225.1 | - | 1 | 360 | ||

Gm11412
Genomic Map