siRNA Details
| Gene Symbol: Gm15764 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC046487.1 | - | 1 | 91 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BE914768.1 | - | 1 | 71 | Chr. 16 - 18416702 - 18421983 [+] on Build GRCm38 | |

Gm15764
Genomic Map