siRNA Details
| Gene Symbol: Gm14635 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045321.1 | 3 | 260 | |||
| GenBank mRNA | AK075658.1 | - | 3 | 261 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm14635| Gene Symbol: Gm14635 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045321.1 | 3 | 260 | |||
| GenBank mRNA | AK075658.1 | - | 3 | 261 | ||