siRNA Details
Gene Symbol: Gm14635 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045321.1 | 4 | 970 | |||
GenBank mRNA | AK075658.1 | - | 4 | 972 | ||
DV655940.1 | - | 1 | 268 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gm14635 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045321.1 | 4 | 970 | |||
GenBank mRNA | AK075658.1 | - | 4 | 972 | ||
DV655940.1 | - | 1 | 268 |