siRNA Details
| Gene Symbol: Gm11715 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | C88942.1 | - | 2 | 201 | ||
| C89518.1 | - | 2 | 215 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm11715| Gene Symbol: Gm11715 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | C88942.1 | - | 2 | 201 | ||
| C89518.1 | - | 2 | 215 | |||