siRNA Details
| Gene Symbol: Uchl1os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_102714.1 | 5 | 1616 | |||
| GenBank mRNA | AK078321.1 | - | 5 | 1617 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Uchl1os| Gene Symbol: Uchl1os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_102714.1 | 5 | 1616 | |||
| GenBank mRNA | AK078321.1 | - | 5 | 1617 | ||