siRNA Details
| Gene Symbol: 9230111E07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK020334.1 | - | 1 | 347 | ||
| BY721144.1 | - | 1 | 347 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

9230111E07Rik| Gene Symbol: 9230111E07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK020334.1 | - | 1 | 347 | ||
| BY721144.1 | - | 1 | 347 | |||