siRNA Details
| Gene Symbol: Gm14703 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK035368.1 | - | 1234 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm14703| Gene Symbol: Gm14703 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK035368.1 | - | 1234 | |||