siRNA Details
| Gene Symbol: 4930447M23Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK076779.1 | - | 1 | 753 | ||
| BY727172.1 | - | 1 | 231 | |||
| CJ050344.1 | - | 1 | 196 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930447M23Rik| Gene Symbol: 4930447M23Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK076779.1 | - | 1 | 753 | ||
| BY727172.1 | - | 1 | 231 | |||
| CJ050344.1 | - | 1 | 196 | |||