siRNA Details
| Gene Symbol: Gm11762 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045099.1 | 3 | 1327 | |||
| GenBank mRNA | AK053782.1 | - | 3 | 1330 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB546098.2 | - | 3 | 306 | Chr. 11 - 119549712 - 119569042 [+] on Build GRCm38 | |

Gm11762
Genomic Map