siRNA Details
Gene Symbol: C330006A16Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK077073.1 | - | 1 | 1249 | ||
AK082757.1 | - | 1 | 1252 | |||
BC020361.1 | - | 1 | 1190 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: C330006A16Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK077073.1 | - | 1 | 1249 | ||
AK082757.1 | - | 1 | 1252 | |||
BC020361.1 | - | 1 | 1190 |