siRNA Details
| Gene Symbol: 1200007C13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK004627.1 | - | 2 | 1475 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1200007C13Rik| Gene Symbol: 1200007C13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK004627.1 | - | 2 | 1475 | ||