siRNA Details
| Gene Symbol: Gm16291 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045788.1 | 1 | 92 | |||
| GenBank mRNA | AK038507.1 | - | 1 | 93 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB632278.1 | - | 1 | 93 | Chr. 15 - 39797045 - 39810428 [+] on Build GRCm38 | |

Gm16291
Genomic Map