siRNA Details
| Gene Symbol: 1700007P06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040554.1 | 2 | 199 | |||
| GenBank mRNA | AK005744.1 | - | 2 | 200 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CN836487.1 | - | 2 | 202 | Chr. 1 - 187125138 - 187125917 [+] on Build GRCm38 | |

1700007P06Rik
Genomic Map