siRNA Details
| Gene Symbol: Gm38691 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK076930.1 | - | 2 | 562 | ||
| Gene Symbol: Gm16226 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AV206142.2 | - | 514 | |||
| AV209032.2 | - | 494 | ||||

Gm38691
Genomic Map