siRNA Details
| Gene Symbol: 4930556A20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK016140.1 | - | 1 | 94 | ||
| Gene Symbol: Gm16555 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK132896.1 | - | 1 | 83 | ||

4930556A20Rik
Genomic Map