siRNA Details
| Gene Symbol: Gm1965 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_121593.1 | 3 | 1126 | |||
| NR_121594.1 | 3 | 1112 | ||||
| NR_121595.1 | 3 | 1061 | ||||
| NR_121596.1 | 2 | 997 | ||||
| GenBank mRNA | AK136244.1 | - | 2 | 999 | ||
| AK139519.1 | - | 3 | 1115 | |||
| AK165099.1 | - | 3 | 1099 | |||

Gm1965
Genomic Map