siRNA Details
| Gene Symbol: Gm4524 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK045496.1 | - | 2 | 411 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB643919.1 | - | 2 | 411 | Chr. 16 - 25956475 - 25958868 [+] on Build GRCm38 | |

Gm4524
Genomic Map