siRNA Details
| Gene Symbol: 9130221H12Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046001.1 | 1 | 295 | |||
| GenBank mRNA | BC022632.1 | - | 2 | 225 | 4207058 | |
| Gene Symbol: Zfp526 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK173308.1 | - | 1479 | |||

9130221H12Rik
Genomic Map