siRNA Details
| Gene Symbol: C030017D09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK021088.1 | - | 2 | 650 | ||
| AK047727.1 | - | 2 | 652 | |||
| BB284396.1 | - | 1 | 194 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C030017D09Rik| Gene Symbol: C030017D09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK021088.1 | - | 2 | 650 | ||
| AK047727.1 | - | 2 | 652 | |||
| BB284396.1 | - | 1 | 194 | |||