siRNA Details
| Gene Symbol: Gm17066 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040628.1 | 1 | 531 | |||
| GenBank mRNA | AK005802.1 | - | 1 | 532 | ||
| CK334289.1 | - | 1 | 316 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm17066| Gene Symbol: Gm17066 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040628.1 | 1 | 531 | |||
| GenBank mRNA | AK005802.1 | - | 1 | 532 | ||
| CK334289.1 | - | 1 | 316 | |||