siRNA Details
Gene Symbol: Gm17066 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040628.1 | 1 | 504 | |||
GenBank mRNA | AK005802.1 | - | 1 | 505 | ||
CK334289.1 | - | 1 | 289 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gm17066 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040628.1 | 1 | 504 | |||
GenBank mRNA | AK005802.1 | - | 1 | 505 | ||
CK334289.1 | - | 1 | 289 |