siRNA Details
| Gene Symbol: 1700116B05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK018963.1 | - | 2 | 366 | ||
| BY707292.1 | - | 2 | 363 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700116B05Rik| Gene Symbol: 1700116B05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK018963.1 | - | 2 | 366 | ||
| BY707292.1 | - | 2 | 363 | |||