siRNA Details
| Gene Symbol: Malat1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002847.2 | 1 | 5906 | |||
| GenBank mRNA | AK141413.1 | - | 1 | 990 | ||
| EF177380.1 | - | 1 | 5906 | |||
| FJ209304.1 | - | 1 | 5906 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Malat1| Gene Symbol: Malat1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002847.2 | 1 | 5906 | |||
| GenBank mRNA | AK141413.1 | - | 1 | 990 | ||
| EF177380.1 | - | 1 | 5906 | |||
| FJ209304.1 | - | 1 | 5906 | |||