siRNA Details
| Gene Symbol: A930015D03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015618.2 | 2 | 145 | |||
| GenBank mRNA | AK020860.1 | - | 2 | 146 | ||
| AK078380.1 | - | 2 | 196 | |||
| AK142942.1 | - | 2 | 141 | |||
| BC055918.1 | - | 2 | 123 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

A930015D03Rik| Gene Symbol: A930015D03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015618.2 | 2 | 145 | |||
| GenBank mRNA | AK020860.1 | - | 2 | 146 | ||
| AK078380.1 | - | 2 | 196 | |||
| AK142942.1 | - | 2 | 141 | |||
| BC055918.1 | - | 2 | 123 | |||