siRNA Details
| Gene Symbol: Erich3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK005945.1 | - | 2 | 978 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Erich3| Gene Symbol: Erich3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK005945.1 | - | 2 | 978 | ||