siRNA Details
| Gene Symbol: 2210019I11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038157.1 | 2, 3 | 425 | |||
| GenBank mRNA | AK008755.1 | - | 2, 3 | 425 | ||
| AK133519.1 | - | 2, 3 | 250 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

2210019I11Rik| Gene Symbol: 2210019I11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038157.1 | 2, 3 | 425 | |||
| GenBank mRNA | AK008755.1 | - | 2, 3 | 425 | ||
| AK133519.1 | - | 2, 3 | 250 | |||