siRNA Details
| Gene Symbol: Rab26os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045289.1 | 1 | 43 | |||
| GenBank mRNA | AK011460.1 | - | 1 | 45 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rab26os| Gene Symbol: Rab26os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045289.1 | 1 | 43 | |||
| GenBank mRNA | AK011460.1 | - | 1 | 45 | ||