siRNA Details
| Gene Symbol: 1700055C04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040726.1 | 3 | 272 | |||
| GenBank mRNA | AK006792.1 | - | 3 | 273 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700055C04Rik| Gene Symbol: 1700055C04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040726.1 | 3 | 272 | |||
| GenBank mRNA | AK006792.1 | - | 3 | 273 | ||