siRNA Details
| Gene Symbol: 4933438K21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045446.1 | 3 | 673 | |||
| GenBank mRNA | AK017111.1 | - | 3 | 674 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4933438K21Rik| Gene Symbol: 4933438K21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045446.1 | 3 | 673 | |||
| GenBank mRNA | AK017111.1 | - | 3 | 674 | ||