siRNA Details
| Gene Symbol: Gm9866 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045868.1 | 4 | 1550 | |||
| NR_045869.1 | 3 | 1175 | ||||
| NR_045870.1 | 2 | 1117 | ||||
| GenBank mRNA | AK044503.1 | - | 2 | 1239 | ||
| AK163447.1 | - | 4 | 1346 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm9866| Gene Symbol: Gm9866 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045868.1 | 4 | 1550 | |||
| NR_045869.1 | 3 | 1175 | ||||
| NR_045870.1 | 2 | 1117 | ||||
| GenBank mRNA | AK044503.1 | - | 2 | 1239 | ||
| AK163447.1 | - | 4 | 1346 | |||