siRNA Details
| Gene Symbol: Gm9866 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045868.1 | 4 | 1142 | |||
| NR_045869.1 | 3 | 767 | ||||
| NR_045870.1 | 2 | 709 | ||||
| GenBank mRNA | AK044503.1 | - | 2 | 831 | ||
| AK163447.1 | - | 4 | 938 | |||
| BE648300.1 | - | 2 | 461 | |||

Gm9866
Genomic Map