siRNA Details
| Gene Symbol: Gm17308 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK142469.1 | - | 149 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB660652.1 | - | 1 | 147 | Chr. 3 - 5090277 - 5093327 [+] on Build GRCm38 | |

Gm17308
Genomic Map